Learn More
Brady™ BBP™ Media Wiper
Click to view available options
Color:
Yellow
Unit Size:
1 pack
Description
- Package includes five media wiper pads (printer sold separately)
- For use with BBP™30, BBP™31, and BBP™33, label printers
- Essential for keeping the label media clean and free of debris
- Cleans the label tape before it passes through the thermal transfer print head
Specifications
Specifications
| Product Type | Media Wiping Blade |
| Color | Yellow |
| For Use With (Equipment) | BBP30 Label Printer, BBP31 Sign and Label Printer, BBP33 Label Printer, BBP35 Multi-Color Sign and Label Printer, BBP37 Color and Cut Sign and Label Printer |
| Width (English) | 1.25 in. |
| Height (English) | 5.15 in. |
| Height (Metric) | 13.08 cm |
| Depth (English) | 0.6 in. |
| Depth (Metric) | 1.52 cm |
| Sterility | Non-sterile |
| Format | Tape |
| Show More |