Learn More
Brady™ BBP™ Cutter Cleaning Tool
Product Code. 11736645 Shop All Brady Products
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11736645 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11736645 Supplier Brady™ Supplier No. 142116
  • ONLINE EXCLUSIVE PRICE
    35.00€
    31.60€ / Pack of 1
    Save 3.40€
    9% Off
Estimated Shipment: 10-11-2026
Use the BBP™ Cutter Cleaning Tool to scrape residue off the cutting blade while cleaning your B30 Series Label Printer.
  • Package includes Cutter Cleaning Tool only (printer sold separately)
  • For use with B30 Series Label Printers, including the BBP™31 and BBP™33 Label Printers
  • Clean the printer cutter regularly to keep your label printer at optimal performance levels
  • Use a printer cleaning kit (not included) that includes Isopropyl Alcohol soaked soft foam swabs to wipe away any remaining residue
Specifications
Product Type Cutter Tool
Color Blue
For Use With (Equipment) BBP31 Sign and Label Printer
Sterility Non-sterile
Includes 1 Tool
For Use With (Application) Full ranges of labels for cable and wire ID, facility and safety ID, product ID and circuit board ID
Quantity 1 Ea.
Alternate Part Number B31-CCT