Learn More
Brady™ BBP™ Blank White Pre-Cut Blank Sign Headers
Product Code. 11767254
Click to view available options
Dimensions (LxW):
2.9 H x 7.2 cm W
4 x 6 in.
5.7 H x 7.6 cm W
5.7 H x 9.8 cm W
Quantity:
175/Pk.
250/Pk.
300/Pk.
Unit Size:
Each
Product Code. 11767254
Brand: Brady™ 142194
BBP™ Blank White Pre-Cut Blank Sign Headers are made of durable, low-shrink vinyl with ultra-aggressive adhesive that is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
  • Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
  • 8-10 year average outdoor durability in temperatures from -40°F to 180°F (-40°C to 82°C); can be applied as low as 0°F (-18°C)
Specifications
Certifications/Compliance ASME ANSI Z535, OSHA 1910.145
Description Outdoor Grade Vinyl Labels for BBP3x Printers
Adhesive Type Permanent Acrylic
Color White
Finish Gloss
Material Vinyl
Product Type Pre-cut Blank Label
For Use With (Application) Pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
For Use With (Equipment) BBP30, BBP31, BBP33, BBP35, and BBP37 printers
Quantity 175/Pk.
Temperature Range (English) -40°F to +180 °F
Temperature Range (Metric) -40°C to +82°C
Thickness (English) 0.004 in.
Dimensions (LxW) 4 x 6 in.
Width (English) 6 in.
Width (Metric) 152.4 mm
Height (English) 4 in.
Height (Metric) 152 mm
Alternate Part Number B30-25-595-BLNKWT