Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- GC Columns
- GC Capillary Columns
- Trajan™ BPX5 GC Capillary Columns
Trajan™ BPX5 GC Capillary Columns
Product Code. 10208670
Change view
Click to view available options
Length (Metric):
6 m
10 m
12 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.15 μm
0.25 μm
0.5 μm
1 μm
Unit Size:
Each
30 product options available for selection
| Product Code. | Length (Metric) | Diameter (Metric) Inner | Film Thickness | unitSize |
|---|---|---|---|---|
| 10208670 | 60 m | 0.32 mm | 0.25 μm | Each |
| 15218653 | 6 m | 0.32 mm | 1 μm | Each |
| 10567342 | 12 m | 0.22 mm | 0.25 μm | Each |
| 10442341 | 25 m | 0.32 mm | 0.25 μm | Each |
| 10519452 | 12 m | 0.53 mm | 0.25 μm | Each |
| 12684945 | 60 m | 0.25 mm | 0.25 μm | Each |
| 16662612 | 60 m | 0.32 mm | 1 μm | Each |
| 16642612 | 30 m | 0.53 mm | 1 μm | Each |
| 16622612 | 25 m | 0.53 mm | 1 μm | Each |
| 10320941 | 50 m | 0.22 mm | 0.25 μm | Each |
| 10269190 | 25 m | 0.53 mm | 0.25 μm | Each |
| 15124079 | 30 m | 0.53 mm | 0.5 μm | Each |
| 10430081 | 25 m | 0.32 mm | 0.5 μm | Each |
| 10268810 | 25 m | 0.22 mm | 0.25 μm | Each |
| 10451031 | 25 m | 0.15 mm | 0.25 μm | Each |
| 10791412 | 30 m | 0.22 mm | 0.25 μm | Each |
| 10567532 | 12 m | 0.32 mm | 0.25 μm | Each |
| 10025450 | 12 m | 0.53 mm | 1 μm | Each |
| 10300711 | 12 m | 0.32 mm | 1 μm | Each |
| 15288643 | 15 m | 0.25 mm | 1 μm | Each |
| 10616021 | 30 m | 0.32 mm | 0.25 μm | Each |
| 10177200 | 15 m | 0.32 mm | 0.25 μm | Each |
| 12905561 | 15 m | 0.25 mm | 0.25 μm | Each |
| 10762562 | 50 m | 0.32 mm | 0.5 μm | Each |
| 15298643 | 30 m | 0.25 mm | 1 μm | Each |
| 10661813 | 10 m | 0.1 mm | 0.1 μm | Each |
| 10055400 | 50 m | 0.32 mm | 1 μm | Each |
| 15248643 | 30 m | 0.15 mm | 0.15 μm | Each |
| 15238643 | 30 m | 0.25 mm | 0.1 μm | Each |
| 15228663 | 15 m | 0.25 mm | 0.1 μm | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Very low bleed
- High temperature
- Non-polar
- Ideal for GCMS
- Applications include pesticides, herbicides, hydrocarbons, amines and phenols
Specifications
Specifications
| Max. Temperature | 370°C |
| Functionality/Interaction Type | Pi Bonding with the Aromatic Phenyl Groups |
| Diameter (Metric) Inner | 0.32 mm |
| Stationary Phase | 5% Phenyl Polysilphenylene Siloxane |
| USP Type | G27, G36, G41 |
| For Use With (Application) | Phenols Analysis, Pesticides and Herbicides Analysis, Hydrocarbons, Solvents Analysis, Phenols, Amines, GC/MS |
| Temperature Range | -40°C to +360/370°C |
| Film Thickness | 0.25 μm |
| Length (Metric) | 60 m |
| Packing Material | BPX5 |
| Polarity | Non polar |
| Coating | Fused Silica |
| Show More Show Less |