Learn More
Mutexil™ Charlotte Disposable Round Caps
Click to view available options
Diameter (Metric):
24 cm
48 cm
53 cm
Product Type:
Bouffant
Quantity:
1000/Pk.
500/Pk.
Unit Size:
Pack of 1000
Pack of 500
Description
- Round
- Polypropylene
- Non-woven
- White
Specifications
Specifications
| Product Type | Bouffant |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Quantity | 1000/Pk. |
| Diameter (Metric) | 24 cm |