Learn More
Retsch™ Bottom Sieves SK 300 with Round Holes
Retsch™ Bottom Sieves SK 300 with Round Holes are constructed of stainless steel for durability and suitable for use with Cross Beater Mill SK 300.
Brand: Retsch™ 02.407.0110
Additional Details : Weight : 0.10000kg
Description
- Stainless steel
- Round holes
- For use with cross beater
Specifications
Round holes | |
Sieve | |
Cross Beater Mill SK 300 |
8.00 mm | |
Stainless Steel |