Learn More
Retsch™ Bottom Sieves SK 300 with Round Holes
Retsch™ Bottom Sieves SK 300 with Round Holes are constructed of stainless steel for durability and suitable for use with Cross Beater Mill SK 300.
231.00€
Specifications
Description | Round holes |
---|---|
Type | Sieve |
Material | Stainless Steel |
For Use With (Equipment) | Cross Beater Mill SK 300 |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
15899254
|
Retsch™
02.407.0106 |
3.00 mm |
231.00€
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
15809264
|
Retsch™
02.407.0107 |
4.00 mm |
231.00€
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
15819264
|
Retsch™
02.407.0108 |
5.00 mm |
231.00€
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
15829264
|
Retsch™
02.407.0109 |
6.00 mm |
231.00€
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
15839264
|
Retsch™
02.407.0110 |
8.00 mm |
231.00€
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
15849264
|
Retsch™
02.407.0111 |
10.00 mm |
231.00€
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
Description
- Stainless steel
- Round holes
- For use with cross beater
Specifications
Round holes | |
Sieve | |
Stainless Steel | |
Cross Beater Mill SK 300 |