Learn More
Retsch™ Bottom Sieves SK 300 with Round Holes
Retsch™ Bottom Sieves SK 300 with Round Holes are constructed of stainless steel for durability and suitable for use with Cross Beater Mill SK 300.
Brand:  Retsch™ 02.407.0109
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15829264
  • 231.00€ / Each
Estimated Shipment: 08-07-2024
Description
  • Stainless steel
  • Round holes
  • For use with cross beater
Specifications
Specifications
Round holes
Sieve
Cross Beater Mill SK 300
6.00 mm