Learn More
Karl Hecht™ Bottle, Narrow Neck with Glass Stopper
Click to view available options
Capacity (Metric):
50 mL
250 mL
500 mL
Color:
Amber
Clear
Unit Size:
Each
Description
- Narrow neck.
- Round shoulder.
- Ground in flat glass stopper.
- Available in clear glass or amber glass.
Specifications
Specifications
| Closure Material | Glass |
| Color | Amber |
| Capacity (Metric) | 250 mL |
| Material | Glass |
| Neck Style | Narrow |
| Product Type | Bottle |
| Shape | Round Shoulder |
| Closure Type | Stopper |