Learn More
B2M™ Bottle Brushes
Click to view available options
Length (Metric) Handle:
235 mm
250 mm
260 mm
270 mm
300 mm
400 mm
450 mm
500 mm
Length (Metric) Brush:
50 mm
75 mm
90 mm
100 mm
110 mm
120 mm
Diameter (Metric) Bristle:
4 mm
8 mm
10 mm
13 mm
15 mm
18 mm
20 mm
25 mm
50 mm
55 mm
Tip Type:
2 Wicks
Bound Bristle Tip
Cotton Tip
Return End
Shell End
Without End
Unit Size:
Pack of 10
Description
- Three choices of bristles:
- White silk bristles for general cleaning use
- Nylon bristles are very resistant for use with aggressive substances
- Goat hair bristles are very soft for delicate cleaning
- A variety of tip styles, including: bound-bristle, 2 wicks, cotton and shell tips
- For cleaning all receptacles: tubes, burettes, balls and bottles
Specifications
Specifications
| Product Type | Bottle Brush |
| Bristle Material | Nylon |
| Tip Type | Shell End |
| Diameter (Metric) Bristle | 50 mm |
| For Use With (Application) | General purpose cleaning, With aggressive substances |
| For Use With (Equipment) | Flacon bottle |
| Length (Metric) Brush | 100 mm |
| Length (Metric) Handle | 270 mm |
| Quantity | 10 Pk. |