Learn More
Bosentan Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Bosentan Monohydrate |
| CAS | 157212-55-0 |
| Molecular Formula | C27 H29 N5 O6 S . H2 O |
| Quantity | 50 mg |
| Synonym | Benzenesulfonamide, 4-(1,1-dimethylethyl)-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)[2,2'-bipyrimidin]-4-yl]-, hydrate (1:1), Benzenesulfonamide, 4-(1,1-dimethylethyl)-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)[2,2'-bipyrimidin]-4-yl]-, monohydrate (9CI), Bosentan hydrate, Bosentan monohydrate |
| InChI Formula | InChI=1S/C27H29N5O6S.H2O/c1-27(2,3)18-10-12-19(13-11-18)39(34,35)32-23-22(38-21-9-6-5-8-20(21)36-4)26(37-17-16-33)31-25(30-23)24-28-14-7-15-29-24;/h5-15,33H,16-17H2,1-4H3,(H,30,31,32);1H2 |
| SMILES | O.COc1ccccc1Oc2c(NS(=O)(=O)c3ccc(cc3)C(C)(C)C)nc(nc2OCCO)c4ncccn4 |
| IUPAC Name | 4-tert-butyl-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidin-4-yl]benzenesulfonamide;hydrate |
| Molecular Weight (g/mol) | 569.63 |
| Formula Weight | 569.1944 |