Learn More
Thermo Scientific BOROSILICATE GLASS 250ml
Brand:  Thermo Scientific 76001135
BOROSILICATE GLASS 250ml
Product Code. 10217696
  • ONLINE EXCLUSIVE PRICE
    83.25€
    79.60€ / Each
    Save 3.65€
    4% Off
Only null left
Estimated Shipment: 08-04-2026