Learn More
Karl Hecht™ Assistent™ Glass Weighing Boats
Product Code. p-8007118
Click to view available options
Capacity (Metric):
3 mL
6 mL
10 mL
Unit Size:
Each
Product Code. 10489621
Brand: Karl Hecht™ 44248010
Only null left
Add to basket
Add to basket
For easy transfer of liquids and powders in the laboratory. Karl Hecht™ Assistent™ Glass Weighing Boats are manufactured from durable DURAN™ borosilicate 3.3 glass.
  • Available in different capacities
  • Clear glass
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 10 mL
Material Borosilicate Glass
Height (Metric) 105 mm
Product Type Weighing Funnel