Learn More
Karl Hecht™ Assistent™ Glass Weighing Boats
Product Code. 10489621
Change view
Click to view available options
Capacity (Metric):
3 mL
6 mL
10 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10489621 10 mL Each
10419061 6 mL Each
10268291 3 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10489621 Supplier Karl Hecht™ Supplier No. 44248010
  • ONLINE EXCLUSIVE PRICE
    19.50€
    11.61€ / Pack of 1
    Save 7.89€
    40% Off
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
For easy transfer of liquids and powders in the laboratory. Karl Hecht™ Assistent™ Glass Weighing Boats are manufactured from durable DURAN™ borosilicate 3.3 glass.
  • Available in different capacities
  • Clear glass
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 10 mL
Material Borosilicate Glass
Height (Metric) 105 mm
Product Type Weighing Funnel