Learn More
Rogo Sampaic™ Borosilicate Glass Test Tubes
Click to view available options
Diameter (Metric):
12 mm
16 mm
20 mm
Length (Metric):
120 mm
160 mm
200 mm
Unit Size:
Pack of 100
Description
- Corrosion-free and resistant to thermal shock
- Autoclavable
- Mouth slightly annealed and rounded
Specifications
Specifications
| Length (Metric) | 120 mm |
| Autoclavable | Autoclavable |
| Diameter (Metric) | 12 mm |