Learn More
Fisherbrand™ Borosilicate Glass Test Tube
Product Code. p-8002026 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
75 mm
100 mm
125 mm
150 mm
Diameter (Metric):
12 mm
16 mm
24 mm
Unit Size:
Pack of 100
Pack of 50
Product Code. 11932208
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Store test reagents and materials. Fisherbrand™ borosilicate glass test tubes come in a variety of sizes.
  • Borosilicate glass test tubes, grade 3.3
  • All ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 4142
Length (Metric) 100 mm
Material Borosilicate Glass
Quantity 100 Pack
Diameter (Metric) 12 mm