Learn More
Fisherbrand™ Borosilicate Glass Test Tube
Product Code. 11992208 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
75 mm
100 mm
125 mm
150 mm
Diameter (Metric):
12 mm
16 mm
24 mm
Unit Size:
Pack of 100
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
11992208 125 mm 16 mm Pack of 100
11932208 100 mm 12 mm Pack of 100
11942198 75 mm 12 mm Pack of 100
11992178 150 mm 24 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11992208
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Store test reagents and materials. Fisherbrand™ borosilicate glass test tubes come in a variety of sizes.
  • Borosilicate glass test tubes, grade 3.3
  • All ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 4142
Length (Metric) 125 mm
Material Borosilicate Glass
Diameter (Metric) 16 mm