Learn More
Witeg™ Borosilicate Glass Tall Form Beakers
Product Code. 11867850
Click to view available options
Capacity (Metric):
250 mL
1000 mL
2000 mL
Height (Metric):
117 mm
120 mm
180 mm
240 mm
Unit Size:
Pack of 10
Pack of 2
Product Code. 11867850
Brand: Witeg™ 5 502 250 S
Only null left
Add to basket
Add to basket
Witeg™ Borosilicate Glass Tall Form Beakers conveniently include graduations and a pouring spout.
  • Durable borosilicate glass
  • Graduated
  • Tall form with spout
  • Some models include handles
Specifications
Diameter (Metric) Outer 61 mm
Capacity (Metric) 250 mL
Material Borosilicate Glass (3.3)
Height (Metric) 117 mm
Includes Spout
Quantity 10 Pk.
Type Beaker
Graduated Yes
Certifications/Compliance ISO 3819, DIN 12331
Style Tall Form