Learn More
Borosilicate Glass T-shape Stopcock Adapter
Click to view available options
Port Size:
14/23
19/26
24/29
Unit Size:
Each
Description
- With GP Rotaflo™ PTFE stopcock providing fine flow control and positive closure
- Detachable plastic screw thread connectors allow rubber hose etc. , to be fitted easily and safely
Specifications
Specifications
| Material | Borosilicate Glass |
| Port Size | 14/23 |
| Shape | T-shape |
| Includes | GP Rotaflo™ PTFE stopcock |
| Product Type | Stopcock Adapter |