Learn More
Borosilicate Glass Stopcock Adapter with Cone
Click to view available options
Port Size:
14/23
19/26
24/29
29/32
Unit Size:
Each
Description
Control flow rate. The QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
- With GP Rotaflo™ PTFE stopcock: - No lubrication required - No contaminating greases - Fine flow control - Positive closure
- Detachable plastic screw thread connectors allow rubber hose etc. , to be fitted easily and safely
Specifications
Specifications
| Material | Borosilicate Glass |
| Port Size | 19/26 |
| Includes | Cone, screw thread adapter and GP Rotaflo™ PTFE stopcock |
| Product Type | Stopcock Adapter |