Learn More
GPE Scientific™ Borosilicate Glass Stirrer
Click to view available options
Standard Taper Joint:
B19
B24
B29
Unit Size:
Each
Description
- For use with overhead stirrer motors
- Non-whip and autoclavable
- The PTFE blades are easily removed or replaced
Specifications
Specifications
| For Use With (Equipment) | Overhead Stirrer Motors |
| Length (Metric) | 465 mm |
| Standard Taper Joint | B29 |
| Type | Uni-Form Precision Glass Stirrers |
| Material | Borosilicate Glass |