Learn More
Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug
Description
- Without scale
- With NS socket and PE stopper
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Material | Borosilicate Glass |
| Quantity | 2Pack |
| Stopper Material | Polyethylene |