Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug and Graduations
Product Code. 10018360
Click to view available options
Capacity (Metric):
50 mL
500 mL
1000 mL
missing translation for 'unitSize'
Pack of 2
Product Code. 10018360
missing translation for 'mfr': Lenz Laborglasinstrumente™ 04022070
Only null left
Add to basket
Add to basket
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug and Graduations are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
  • Cylindrical shape with straight discharge tube
  • With NS socket and PE stopper
  • With solid glass plug
  • With Screwthreade retaining device
  • With scale
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Graduated Yes
Includes Solid Glass plug and screw thread retaining device
Graduations 20mL
Joint Number 29/32 NS Socket
Product Type Separatory Funnel