Learn More
Cole-Parmer™ Stuart™ Borosilicate Glass Receiving Flask
Product Code. 12722375
Change view
Click to view available options
Volume (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12722375 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12722375 Supplier Cole-Parmer™ Supplier No. 1009670
  • ONLINE EXCLUSIVE PRICE
    130.00€
    114.89€ / Pack of 1
    Save 15.11€
    11% Off
Estimated Shipment: 14-01-2027
Specifications
Material Borosilicate Glass
Type Receiving Flask
For Use With (Equipment) For RE100 Rotary Evaporator
Quantity 1 Each
Shape Round
Volume (Metric) 250 mL