Learn More
Fisherbrand™ Borosilicate Glass Petri Dishes
Product Code. 12931408 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric):
60 mm
80 mm
100 mm
120 mm
150 mm
Height (Metric):
12 mm
15 mm
20 mm
25 mm
Unit Size:
Each
Pack of 18
Pack of 20
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Height (Metric) Unit Size
12931408 120 mm 20 mm Pack of 18
16190741 100 mm 15 mm Pack of 20
12921408 100 mm 15 mm Pack of 18
16120751 60 mm 12 mm Pack of 20
12901408 60 mm 12 mm Pack of 18
16130751 120 mm 20 mm Pack of 20
16110751 150 mm 25 mm Pack of 20
12941408 150 mm 25 mm Each
12911408 80 mm 15 mm Pack of 18
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12931408
Estimated Shipment: 23-12-2026
High-quality petri dish, with lid.
  • Constructed of soda lime glass.
  • Includes lid
Specifications
Closure Included Closure Included
Material Soda Lime Glass
Shape Round
Type Petri Dish
Height (Metric) 20 mm
Diameter (Metric) 120 mm