Learn More
Fisherbrand™ Borosilicate Glass Narrow Neck Erlenmeyer Flasks With Pouring Rim and Labeling Area
Product Code. 11932243 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
300 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11932243 300 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11932243
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Measure precise amounts of media and pour them neatly with the Fisherbrand™Borosilicate Glass Narrow Neck Erlenmeyer Flasks With Pouring Rim and Labeling Area.
  • Erlenmeyer
  • Graduated.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN ISO 1773
Capacity (Metric) 300 mL
Material Borosilicate Glass
Height (Metric) 156 mm
Mouth Style Narrow
Diameter (Metric) Outer 50 mm