Learn More
Kimble Chase™ Borosilicate Glass Gas Sampling Bulbs
Product Code. 11760409
Change view
Click to view available options
Product Type:
Volume: 125 mL
Volume: 250 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11760409 Volume: 250 mL Each
11740409 Volume: 125 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11760409 Supplier Kimble Chase™ Supplier No. 6531500250
  • ONLINE EXCLUSIVE PRICE
    244.00€
    225.16€ / Pack of 1
    Save 18.84€
    7% Off
 Restricted Item
Estimated Shipment: 12-05-2027