Learn More
Quickfit™ Borosilicate Glass Digestion Tube with Socket
Click to view available options
Diameter (Metric) Outer:
16 mm
26 mm
42 mm
Overall Length (Metric):
145 mm
255 mm
300 mm
Unit Size:
Pack of 20
Pack of 5
Description
- Contains Quickfit precision-ground socket to accept air condensers
- Manufactured from borosilicate glass for highest chemical and thermal durability
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Diameter (Metric) Outer | 16 mm |
| Overall Length (Metric) | 145 mm |
| Packaging Quantity | 5/Pk. |
| Product Type | Digestion Tube with Socket |