Learn More
Rogo Sampaic™ Glass Coverslips
Product Code. 11727394
Change view
Click to view available options
Dimensions (L x W):
14 mm Dia.
22 x 32 mm
22 x 40 mm
24 x 24 mm
Material:
Borosilicate Glass
Soda Lime Glass
Unit Size:
Pack of 100
Pack of 200
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Material unitSize
11727394 24 x 24 mm Borosilicate Glass Pack of 200
11747394 22 x 40 mm Borosilicate Glass Pack of 100
11737394 22 x 32 mm Borosilicate Glass Pack of 100
13286718 14 mm Dia. Soda Lime Glass Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11727394 Supplier Rogo Sampaic™ Supplier No. BPD020
Only null left
Add to basket
Edge
Add to basket
Preserve and protect specimens on slides.
  • Available in a range of sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Product Type Coverslip
Shape Square
Material Borosilicate Glass
Length (Metric) 24 mm
Width (Metric) 24 mm
Dimensions (L x W) 24 x 24 mm
Quantity 200/Pk.