Learn More
Marienfeld Superior™ Borosilicate Glass Cover Glasses, Round
Click to view available options
Diameter (Metric):
12 mm
16 mm
18 mm
20 mm
24 mm
25 mm
Unit Size:
Pack of 1000
Description
- Made of chemically resistant borosilicate glass D 263™ M of first hydrolytic class
- For light and fluorescence microscopy, especially in medical and biological science
- Excellent flatness, absolutely colorless, perfectly clear
- Refractive index ne: 1.5255
- Abbe value ve: 55
- In compliance with DIN ISO 8255
- For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark and batch number for comprehensive information and traceability
Specifications
Specifications
| Product Type | Coverslip |
| Shape | Circle |
| Material | Borosilicate Glass D 263 M |
| Thickness (Metric) | #1 (0.13 to 0.16 mm) |
| Diameter (Metric) | 24 mm |
| For Use With (Equipment) | Microscope Slides |
| Certifications | CE Mark |
| Quantity | 1000 |