Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Conical Separating Funnels with Solid Glass Plug, Scale
Product Code. 10604491
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10604491 100 mL Pack of 2
11822672 1000 mL Pack of 2
10664861 250 mL Pack of 2
11836134 2000 mL Each
11812672 500 mL Pack of 2
11832662 50 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10604491 Supplier Lenz Laborglasinstrumente™ Supplier No. 04002037
  • ONLINE EXCLUSIVE PRICE
    83.00€
    64.32€ / Pack of 2
    Save 18.68€
    22% Off
Estimated Shipment: 23-11-2026
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Conical Separating Funnels with Solid Glass Plug, Scale are perfect for liquid-liquid extractions and separating chemical components of a mixture into immiscible solvent phases of different densities.
  • With NS socket and PE stopper
  • with solid glass plug
  • with screwthread retaining device
  • with scale (approx. volume /ml)
  • material: borosilicate glass 3.3
Specifications
Capacity (Metric) 100 mL
Material Borosilicate Glass
Quantity 2Pack
Stopper Material Polyethylene