Learn More
Lenz Laborglasinstrumente™ Separating Funnels with Glass Plug, without Scale
Product Code. 10465601
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10465601 1000 mL Pack of 2
15330895 250 mL Pack of 2
10728731 100 mL Pack of 2
10475601 2000 mL Each
15330885 500 mL Pack of 2
10507551 50 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10465601 Supplier Lenz Laborglasinstrumente™ Supplier No. 04000070
  • ONLINE EXCLUSIVE PRICE
    171.00€
    111.56€ / Pack of 2
    Save 59.44€
    34% Off
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with Glass Plug, without Scale are manufactured from durable DURAN™ borosilicate glass.
  • Without scale; with solid glass plug
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Quantity 2Pack
Stopper Material Polyethylene