Learn More
Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale
Product Code. 10643161
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10643161 100 mL Pack of 2
11852672 1000 mL Pack of 2
11836084 2000 mL Each
10100121 250 mL Pack of 2
10393511 50 mL Pack of 2
11871593 500 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10643161 Supplier Lenz Laborglasinstrumente™ Supplier No. 04006137
  • ONLINE EXCLUSIVE PRICE
    95.00€
    84.44€ / Pack of 2
    Save 10.56€
    11% Off
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale are manufactured from durable DURAN™ borosilicate glass.
  • With scale (approx. volume/mL)
  • With PTFE plug
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 100 mL
Material Borosilicate Glass
Quantity 2Pack
Stopper Material Polyethylene