Learn More
Kimble Chase™ Borosilicate Glass Class A Square Volumetric Flask
Product Code. p-8003544
Change view
Click to view available options
Capacity (Metric):
500 mL
900 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12724497 500 mL Each
11812943 900 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12724497 Supplier Kimble Chase™ Supplier No. 28046500
Designed to take up less area and pack more closely on shelves and in refrigerators or other storage cabinets
  • Square cross-section with a width less than the widest diameter of a typical round flask
  • All sizes cover 25-35% less area than conventional flasks
  • Due to the added glass weight and the square section, flasks also have greater stability
  • Graduation ring is blasted on the neck
  • Calibrated to contain
  • Calibrated to deliver
  • With a marking spot
  • Standard Taper ground glass stopper is supplied with the flask
  • Designed from ASTM Specification E288, Class A requirements
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Certifications/Compliance ASTM Specification E288, Class A
Capacity (Metric) 500 mL
Material Borosilicate Glass
Height (Metric) 320 mm
No. per Case 1
Graduated Yes
Class A
Closure Type Taper stopper
Closure Size 19