Learn More
Kimble Chase™ Borosilicate Glass Chromatography Column with Removable Fritted Disc
Click to view available options
Column Capacity:
75 mL
150 mL
Unit Size:
Each
Specifications
Specifications
| For Use With (Application) | Ion exchange chromatography |
| Quantity | 1 Each |
| Diameter (Metric) Inner | 20 mm |
| Type | Chromatography Column |
| USP Type | Type I |
| Column Capacity | 75 mL |
| Length (Metric) | 250 mm |
| Material | Borosilicate Glass |
| Includes | With removable fritted disc |