Learn More
Kimble Chase™ Borosilicate Glass Chromatography Column with Fritted Disc
Product Code. 11817423
Click to view available options
Length (Metric):
400 mm
Unit Size:
Each
Product Code. 11817423
Brand: Kimble Chase™ 4203200234
Specifications
For Use With (Application) Ion exchange chromatography
Quantity 1 Each
Diameter (Metric) Inner 22 mm
Type Chromatography Column
USP Type Type I
Column Capacity 150 mL
Length (Metric) 400 mm
Material Borosilicate Glass
Includes With fritted disc