Learn More
Kimble Chase™ Borosilicate Glass Chromatography Column with Fritted Disc and PTFE Stopcock Plug
Product Code. 11847433
Click to view available options
Length (Metric):
300 mm
500 mm
600 mm
Diameter (Metric) Inner:
38 mm
50 mm
64 mm
75 mm
Unit Size:
Each
Product Code. 11847433
Brand: Kimble Chase™ 4205400750
Gravity chromatography column manufactured from medium wall tubing with 821000 PTFE stopcock
  • 40-60 micron porosity glass frit at the lower end to act as a support for column packing material
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Ion exchange chromatography
Quantity 1 Each
Diameter (Metric) Inner 75 mm
Type Chromatography Column
USP Type Type I
Column Capacity 2149 mL
Length (Metric) 500 mm
Material Borosilicate Glass
Includes With fritted disc and PTFE stopcock plug