Learn More
Quickfit™ Borosilicate Glass Büchner Vacuum Filtration Funnels with Glass Sintered Disc
Product Code. 15508010
Change view
Click to view available options
Capacity (Metric):
30 mL
80 mL
125 mL
500 mL
Porosity:
1
2
3
4
Diameter (Metric) Disc:
40 mm
40mm
60mm
95 mm
95mm
Disc: 30mm
Unit Size:
Each
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Porosity Diameter (Metric) Disc unitSize
15508010 125 mL 1 60mm Pack of 5
15728127 30 mL 4 Disc: 30mm Pack of 5
15528010 125 mL 4 60mm Pack of 5
15568000 80 mL 1 40mm Pack of 5
15738127 500 mL 2 95mm Pack of 5
15748127 500 mL 4 95mm Pack of 5
14819830 80 mL 4 40mm Each
15598000 80 mL 4 40mm Pack of 5
15588000 80 mL 3 40 mm Pack of 5
15538010 500 mL 3 95 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15508010 Supplier Quickfit Supplier No. VB60/02M
Add to basket
Add to basket
Useful where conventional filter papers would be destroyed by acids and alkalis
  • Manufactured from borosilicate glass for highest resistance to chemical attack
  • With Quickfit ground glass cone to allow direct filtration into Quickfit Büchner flasks
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 125 mL
Material Borosilicate Glass
Height (Metric) 48 mm
Cone Size 24/29
For Use With (Application) For filtering solutions that can be filtered by filter papers which are not resistant to certain acids and bases
Diameter (Metric) Disc 60mm
Porosity 1
Product Type Buchner Vacuum Filtration Funnel