Learn More
Witeg™ Volumetric Flask Class A with ST Blue Graduated
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Pack of 10
Description
- Available with or without stopper
- Borosilicate glass 3.3
- Tolerance according to ASTM E 288
- Batch certificate included
- DE-M marked according to the German calibration law
- Standard and special sizes available
Specifications
Specifications
| Calibration | DE-M marked according to German calibration law |
| Capacity (English) | 0.3 oz. |
| Certifications/Compliance | DIN EN ISO 1042, Tolerance according to ASTM E 288 |
| Closure Material | Polyethylene |
| Color | White |
| Capacity (Metric) | 10 mL |
| Material | Borosilicate Glass (3.3) |
| Includes | Batch certificate |
| Limit of Error | ±0.060 mL |
| Neck Style | Ground Joint |
| Show More |