Learn More
WTW™ Boron B Test Kits
Click to view available options
No. of Tests:
25
Unit Size:
Pack of 25
Specifications
Specifications
| Analytes | Azomethine |
| For Use With (Application) | For waste water, drinking water, sea water |
| For Use With (Equipment) | PhotoLab™ S12, PhotoLab™ spectral photometers |
| Includes | Certificates |
| No. of Tests | 25 |
| Sufficient For | 25 Tests |
| Test Method | Photometric |
| Type | Boron (B) cell test kit |