Learn More
BRAND™ Boro 3.3 Conical Ground Joint Stopper
Product Code. p-7159969 Shop All Brand Products
Click to view available options
Standard Taper Joint:
NS 24/29
NS 45/40
Unit Size:
Each
Product Code. 10597451
Brand: BRAND™ 144530
Only null left
Add to basket
Add to basket
Solid or semi-hollow, with octagonal grip
  • Boro 3.3 glass construction
  • DIN 12252
  • TRUSTED_SUSTAINABILITY
    Specifications
    Description Solid
    Standard Taper Joint NS 24/29
    Material Borosilicate Glass
    Shape Octagonal Grip