Learn More
Tocris Bioscience™ BOP
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
BOP is a dual α9β1/α4β1 integrin inhibitor; rapidly and preferentially mobilizes HSCs and progenitors. Augments HSC mobilization in combination with AMD3100 (Cat. No. 3299).
Fluorogenic, bright and photostable Fluorescent Conjugate BOP-JF646 (Cat. No. 6997) also available.
High affinity fluorescent derivative R-BC154 (Cat. No. 6048) also available.
Specifications
Specifications
| CAS | 1947348-42-6 |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in water and to 100mM in DMSO |
| Molecular Formula | C25H28N3O7SNa |
| Formula Weight | 537.56 |
| Color | White |
| Product Type | Inhibitor |