Learn More
uvex Boots S3
Product Code. 30533585 Shop All Uvex Products
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
Product Code. 30533585
Brand: uvex 6874242
These uvex 3 boots provide reliable protection and comfort in a variety of situations.
The uvex 3S3 68742 safety shoe has been specially developed for the extreme demands faced in heavy applications with SR slip resistance.
  • It combines necessary robustness with a sporty and dynamic design.
  • Equipped with the latest uvex technologies, the uvex 3 reliably protects the wearer while providing high levels of comfort.
  • EN ISO 20345: S3L
TRUSTED_SUSTAINABILITY
Specifications
Color Black, Yellow
Material Leather, Textile
Size 42
Toe Material Polymer
Disposable Reusable
Gender Men's
Fit Standard Width
Latex Free Yes
Waterproof Yes
Product Type Boots