Learn More
Julabo™ Booster pump, magnetically coupled, for PRESTO™ temperature control systems, 2.1 bar
Click to view available options
Type:
Booster Pump
Unit Size:
Each
Description
The Julabo magnetically coupled Booster Pump is the ideal solution to increasing the pressure or flow rate in your application. The Booster Pump is specifically designed to be easily connected between PRESTO™ temperature control units and your application, increasing your fluid pressure by up to 2.1 bar. The stainless steel design of the pump provides excellent resistivity against chemical effects. The magnetically-driven design guarantees 100% leakage free operation over an extraordinary temperature range of -90°C to +250°C.
Suitable fluids include water-glycol, silicone oil, and Fluorinert
Working temperature range: -90°C to 250°CMaximum viscosity: 50 cStMaximum flow rate: 80 L\minPower: 230W at full motor speed
Suitable fluids include water-glycol, silicone oil, and Fluorinert
Working temperature range: -90°C to 250°CMaximum viscosity: 50 cStMaximum flow rate: 80 L\minPower: 230W at full motor speed
Specifications
Specifications
| Type | Booster Pump |