Learn More
Thermo Scientific Chemicals Bone Morphogenetic Protein 2, Human
Product Code. p-5386194
Click to view available options
Quantity:
10μg
2μg
Unit Size:
10µg
2µg
Product Code. 15418709
Brand: Thermo Scientific Chemicals J67332.&E
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For research use
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Lyophilized powder
Quantity 2μg
Source Escherichia coli
Storage Requirements -30° to -10°C
Regulatory Status RUO
Product Type Bone Morphogenetic Protein 2, Human
Recombinant Native
Form Lyophilized