Learn More
Thermo Scientific Chemicals Bone Morphogenetic Protein 2, Human
Click to view available options
Quantity:
10μg
2μg
Unit Size:
10µg
2µg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Formulation | Lyophilized powder |
| Quantity | 2μg |
| Source | Escherichia coli |
| Storage Requirements | -30° to -10°C |
| Regulatory Status | RUO |
| Product Type | Bone Morphogenetic Protein 2, Human |
| Recombinant | Native |
| Form | Lyophilized |