Learn More
Bohlender™ BOLA™ Spare O-rings for Stopcocks
Product Code. 17107474
Click to view available options
Material:
NBR
Unit Size:
Pack of 20
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 17107474
Brand: Bohlender™ J119801
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Spare o-rings for stopcocks.
  • O-rings made of NBR
  • For BOLA™ Eco Stopcock Plugs
  • Taper: 1:10
  • Thread M: 7 mm
  • Suitable for size: NS 10
Specifications
Type Spare O-ring
Material NBR
For Use With (Equipment) For BOLA™ Eco Stopcock Plugs