Learn More
Bohlender™ Screw Caps with Flexible Distributors
Click to view available options
Thread Size (Metric):
8 mm
Unit Size:
Each
Description
- Bent hose connectors on the upper side
- Straight hose connectors on the bottom side
- Restricted chemical resistance
- For working temperatures up to max. +110°C
Specifications
Specifications
| Color | Green |
| For Use With (Application) | Drawing or inserting liquids, for elastic tubing (e.g. Viton™, Tygon™, silicone) |
| Temperature Range (Metric) | -20 to 110 deg.C |
| Thread Size (Metric) | 8 mm |
| Neck Finish | 8 mm |
| Product Type | Screw Cap with Flexible Distributors |
| For Use With (Equipment) | Tubing I.D. 7-11mm |