Learn More
Bohlender™ BOLA™ PTFE Tubing Connectors
Product Code. 10737532
Click to view available options
Tubing Size:
11mm O.D., 6mm I.D.
4.5mm O.D., 2mm I.D.
6.8mm O.D., 3mm I.D.
6.8mm O.D., 5mm I.D.
Unit Size:
Each
Product Code. 10737532
Brand: Bohlender™ D57506
Only null left
Add to basket
Add to basket
Connect elastic tubing with these tubing connectors. BOLA PTFE Tubing Connectors feature universal chemical resistance and are vacuum suitable.
  • PTFE straight fitting
  • Three connectors for elastic tubing (e.g. Viton, Tygon, silicone)
  • Universal chemical resistance
  • Product is only exposed to PTFE
Compliance
FDA
Specifications
Diameter (Metric) Inner 5 mm
Diameter (Metric) Outer 6.8 mm
Overall Length (Metric) 61 mm
Tubing Size 6.8mm O.D., 5mm I.D.