Learn More
Bohlender™ BOLA™ Miniature Luer Connectors, PTFE
Click to view available options
Tubing Size:
0.8 x 1.6mm (I.D. x O.D.)
1.6 x 3.2mm (I.D. x O.D.)
Format:
Female Luer Cone
Male Luer Cone
Unit Size:
Each
Description
- PTFE Miniature Luer Connectors
- For transition from luer connection to female thread UNF 1/4 in. 28 G
- Universal chemical resistance
Compliance
FDA
Specifications
Specifications
| Certifications/Compliance | FDA |
| Description | Luer connector with female luer cone |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 1 mm |
| Material | PTFE |
| For Use With (Application) | Transition from luer connection to female thread UNF 1/4 in. 28 G |
| Min. Operating Temperature (Metric) | -200°C |
| Tubing Size | 0.8 x 1.6mm (I.D. x O.D.) |
| Thread Size | UNF 1/4 in. 28 G |
| Format | Female Luer Cone |
| Show More |